Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers
Replacement Hydrometer for FBA11582 Hydrometer Set.
Manufacturer:  Fisherbrand™ 11583D
 View more versions of this product
Catalog No. 11-583D
Specifications
Description
Hydrometer, Replacement; Fisherbrand; For set of eight precision specific gravity; Division: 0.002; Range: 1.000 to 1.220; Without 3-pt certification
Replacement Hydrometer for FBA11582 Hydrometer Set.
Description & Specifications
Specifications
0.002
Without 3-pt Certification
Set of eight precision specific gravity hydrometer
1 to 1.22
Accessories