Please login to your online account to display your discounted pricing

Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers


Replacement Hydrometer for FBA11582 Hydrometer Set.

Manufacturer: fisherbrand™  11583CC

 View more versions of this product

Catalog No. 11-583CC

  • / Each
 

Description & Specifications

Specifications

Specific Gravity Range 0.9 to 1.01
Certifications/Compliance 3-point Certification
Increments 0.001
For Use With (Equipment) Set of eight precision specific gravity hydrometer

Replacement Hydrometer for FBA11582 Hydrometer Set.