Please login to your online account to display your discounted pricing

Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers


Replacement Hydrometer for FBA11582 Hydrometer Set.

Manufacturer: fisherbrand™  11583B

 View more versions of this product

Catalog No. 11-583B

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Set of eight precision specific gravity hydrometer
Specific Gravity Range 0.795 to 0.91
Increments 0.001
Certifications/Compliance Without 3-pt Certification

Replacement Hydrometer for FBA11582 Hydrometer Set.

Accessories

Accessories for Catalog Number: 11583B

Item Mfr. No. Item Description Price Per Unit Quantity