Please login to your online account to display your discounted pricing

Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers


Replacement Hydrometer for FBA11582 Hydrometer Set.

Manufacturer: fisherbrand™  11583AC

 View more versions of this product

Catalog No. 11-583AC

  • / Each
 

Description & Specifications

Specifications

Specific Gravity Range 0.695 to 0.81
For Use With (Equipment) Set of eight precision specific gravity hydrometer
Certifications/Compliance 3-point Certification
Increments 0.001

Replacement Hydrometer for FBA11582 Hydrometer Set.