Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers
Replacement Hydrometer for FBA11582 Hydrometer Set.
Manufacturer:  Fisherbrand™ 11583GC
 View more versions of this product
Catalog No. 11-583GC
Specifications
Description
Hydrometer, Replacement; Fisherbrand; For set of eight precision specific gravity; Division: 0.002; Range: 1.600 to 1.820; W/3-pt certification
Replacement Hydrometer for FBA11582 Hydrometer Set.
Description & Specifications
Specifications
0.002
3-point Certification
Set of eight precision specific gravity hydrometer
1.6 to 1.82