Please login to your online account to display your discounted pricing

Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers


Replacement Hydrometer for FBA11582 Hydrometer Set.

Manufacturer: fisherbrand™  11583FC

 View more versions of this product

Catalog No. 11-583FC

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Set of eight precision specific gravity hydrometer
Increments 0.002
Specific Gravity Range 1.4 to 1.62
Certifications/Compliance 3-point Certification

Replacement Hydrometer for FBA11582 Hydrometer Set.