Please login to your online account to display your discounted pricing

Fisherbrand™ Replacement Hydrometers for Set of Eight Precision Specific Gravity Hydrometers


Replacement Hydrometer for FBA11582 Hydrometer Set.

Manufacturer: fisherbrand™  11583E

 View more versions of this product

Catalog No. 11-583E

  • / Each
 

Description & Specifications

Specifications

Specific Gravity Range 1.2 to 1.42
Increments 0.002
Certifications/Compliance Without 3-pt Certification
For Use With (Equipment) Set of eight precision specific gravity hydrometer

Replacement Hydrometer for FBA11582 Hydrometer Set.

Accessories

Accessories for Catalog Number: 11583E

Item Mfr. No. Item Description Price Per Unit Quantity  
11-582-1
Thermo Scientific™ Nalgene™ PMP Hydrometer Jar
thermo scientific™
N62300500
Protect against breakage. This PMP jar is lighter than glass and offers superior chemical resistance and a broad base, making it a stable addition to the lab or field. Each for $80.17
Case of 6 Each for $481.02