missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Redi-Tip™ Pipet Tips: Reference

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer:  Fisherbrand™ 112GFIS

 View more versions of this product

Catalog No. 02-681-163



 

Specifications Description & Specifications

Specifications

Redi-Tip™ Tip
Nonsterile
250, 500, 1000μL
Universal Fit
Bulk
6.9cm
Research grade pipetters
101 to 1000μL
Blue
Yes
Polypropylene
2.75 in.
RNase, DNase, DNA, pyrogen-free
Accessories