Please login to your online account to display your discounted pricing

Fisherbrand™ Redi-Tip™ Pipet Tips: Reference


Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer: fisherbrand™  113RGFIS

 View more versions of this product

Catalog No. 02-681-156

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Redi-Tip™ Tip
Volume (Metric) 1 to 200μL
Sterility Nonsterile
Color Yellow
Graduations 10, 50, 100μL
Autoclavable Yes
Tip Style Universal Fit
Material Polypropylene
Packaging Stack Rack, 5 x 200
Length (English) 2 in.
Length (Metric) 5cm
Certifications/Compliance RNase, DNase, DNA, bioburden, PCR inhibitor, pyrogen-free
For Use With (Equipment) Research grade pipetters