Please login to your online account to display your discounted pricing

Fisherbrand™ Redi-Tip™ Pipet Tips: Reference


Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer: fisherbrand™  H11396RGSFIS

 View more versions of this product

Catalog No. 02-681-155

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Packaging Hinged Rack, 10 x 96
Material Polypropylene
Sterility Sterile
For Use With (Equipment) Research grade pipetters
Product Type Redi-Tip™ Tip
Length (English) 2 in.
Length (Metric) 5cm
Autoclavable Yes
Volume (Metric) 1 to 200μL
Certifications/Compliance RNase, DNase, DNA, bioburden, PCR inhibitor, pyrogen-free
Color Yellow
Graduations 10, 50, 100μL
Tip Style Universal Fit