Please login to your online account to display your discounted pricing

Fisherbrand™ Redi-Tip™ Pipet Tips: Reference


Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer: fisherbrand™  10896RGFIS

 View more versions of this product

Catalog No. 02-681-455

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Packaging Lift-off Rack, 10 x 96
Sterility Nonsterile
Material Polypropylene
Product Type Redi-Tip™ Tip
Length (English) 2.31 in.
Length (Metric) 5.87cm
For Use With (Equipment) Research grade pipetters
Volume (Metric) 1 to 300μL
Autoclavable Yes
Tip Style Universal Fit
Graduations 10, 50, 100, 200μL
Color Natural
Certifications/Compliance RNase, DNase, DNA, pyrogen-free