missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Redi-Tip™ Pipet Tips

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer:  Fisherbrand™ 113RGNFIS

 View more versions of this product

Catalog No. 02-681-150



 

Description

Description

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and pyrogen free
  • Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor and pyrogen free

  • Graduations at 10, 50 and 100μL; 10, 50, 100 and 200μL; or 250, 500 and 1000μL
  • Graduations allow for immediate volume verification

Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.

Specifications

Specifications

Universal Fit Pipettor
Universal Fit
Non-sterile
10, 50, 100μL
PP (Polypropylene)
5cm
Universal Fit Pipettor
Rack
1 to 200μL
Natural
Yes
2 in.
DNase, RNase, DNA, and Pyrogen Free
Documents