Please login to your online account to display your discounted pricing

Fisherbrand™ Redi-Tip™ Pipet Tips: General Purpose


Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer: fisherbrand™  H111R1000SFIS

 View more versions of this product

Catalog No. 02-707-509

  • / Pack of 1000 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Redi-Tip™ Tip
Material Polypropylene
Packaging Hinged Rack, 10 x 100
Length (English) 2.75 in.
Length (Metric) 6.9cm
For Use With (Equipment) Research grade pipetters
Sterility Sterile
Autoclavable Yes
Tip Style General Purpose
Certifications/Compliance RNase, DNase, DNA, bioburden, PCR inhibitor, pyrogen-free
Volume (Metric) 101 to 1000μL
Color Blue