Fisherbrand™ Red Washing/Dispensing Bottles
Red for dangerous contents.
$38.79 - $43.10
Specifications
Closure Material Polypropylene
Autoclavable No
Color Red, Red
Material LDPE, Polyethylene
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
03-409-16A Accessories View Documents Fisherbrand™
006720250
250ml Pack of 6 Each for $38.79
Case of 6 Pack for $232.74
03-409-16B Accessories View Documents Fisherbrand™
006720500
500mL Pack of 6 Each for $43.10
Case of 4 Pack for $172.40
Description
Description
  • Polyethylene bottle with integral, one-piece, molded polypropylene cap and nozzle to eliminate leakage
  • Rigid spout permits controlled delivery: single drop or fine stream
  • Tip may be cut back to increase flow
Specifications
Specifications
Polypropylene
No
Red, Red
LDPE, Polyethylene