Please login to your online account to display your discounted pricing

Fisherbrand™ Rectangular Storage Boxes

Made from recyclable cardboard

$136.82 - $378.96

Specifications

Color White
Product Type Sliding Design Storage Box
For Use With (Application) Store a variety of samples such as slides, cassettes, specimens and seeds
Material Recyclable Cardboard
Shape Rectangular
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity    

14-101-A

 
fisherbrand™
F188620001
2.25 x 1.25 x 1.06 in. (5.71 x 3.17 x 2.69cm) Pack of 72 Each for $136.82

14-101-C

 
fisherbrand™
F188620003
3 x 2 x 1.25 in. (7.62 x 5.08 x 3.17cm) Pack of 72 Each for $224.75

14-101-B

 
fisherbrand™
F188620002
2.75 x 1.75 x 1.13 in. (6.98 x 4.44 x 2.87cm) Pack of 72 Each for $146.71

14-101-D

 
fisherbrand™
F188620004
3.63 x 2.5 x 1.5 in. (9.22 x 6.35 x 3.81cm) Pack of 72 Each for $378.96
Description & Specifications

Specifications

Color White
Product Type Sliding Design Storage Box
For Use With (Application) Store a variety of samples such as slides, cassettes, specimens and seeds
Material Recyclable Cardboard
Shape Rectangular

  • Sliding box design
  • Can be used to store a variety of samples such as slides, cassettes, specimens and seeds
  • White interior/exterior for marking or labeling