Please login to your online account to display your discounted pricing

Fisherbrand™ Quantitative-Grade Filter Paper Circles


Limited wet strength.†

Manufacturer: fisherbrand™  097902FEMD

 View more versions of this product

Catalog No. S47577F

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Particle Retention 2 to 5μmm
Format Circle
Grade Q5
For Use With (Application) Widely used for general gravimetric analysis. Retains medium-sized particles such as lead sulfate, calcium oxalate, and magnesium-ammonium
Porosity Medium
Diameter (Metric) 15cm