missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Quantitative-Grade Filter Paper Circles

Limited wet strength.†

Manufacturer:  Fisherbrand™ 097902DEMD

 View more versions of this product

Catalog No. S47577D



Description

Description

Coarse Porosity

  • Flowrate: 2.6 sec./100mL
  • Surface: Smooth, open texture
  • Particle Retention: >10μm
  • Applications: Suitable for coarse and gelatinous precipitates requiring thorough washing on paper
Medium Porosity
  • Flowrate: 15.7 sec./100mL
  • Surface: Smooth
  • Particle Retention: 2 to 5μm
  • Applications: Widely used for general gravimetric analysis. Retains medium-sized particles such as lead sulfate, calcium oxalate, and magnesium-ammonium
Fine Porosity
  • Flowrate: 76 sec./100mL
  • Surface: Smooth, dense texture
  • Particle Retention: >2μm
  • Applications: Excellent retention; suitable for finest precipitates such as metastannic acid, barium sulfate, mercuric chromate and strontium sulfate

Specifications

Specifications

Education
Circle
11cm
Widely used for general gravimetric analysis. Retains medium-sized particles such as lead sulfate, calcium oxalate, and magnesium-ammonium
Q5
Medium
2 to 5μmm
Documents