missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Qualitative-Grade Filter Paper Circles

Limited wet strength.†

$7.35 - $16.55

Specifications

Format Circle
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Grade Porosity Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Grade Porosity Price Quantity  
S47571C Fisherbrand™
09801CEMD
11cm P5 Medium Pack of 100 Each for $9.15
S47573B Fisherbrand™
09795BEMD
7cm P8 Coarse Pack of 100 Each for $12.65
S47573F Fisherbrand™
09795FEMD
15cm P8 Coarse Pack of 100 Each for $13.10
S47574D Fisherbrand™
09801DEMD
12.5cm P5 Medium Pack of 100 Each for $16.55
S47574B Fisherbrand™
09801BEMD
9cm P5 Medium Pack of 100 Each for $7.35
S47574E Fisherbrand™
09801EEMD
15cm P5 Medium Pack of 100 Each for $12.15
Specifications

Description

Coarse Porosity

  • Flowrate: Fast, 175mL/min.
  • Surface: Smooth, open texture
  • Particle Retention: >20μm
  • Applications: Suitable for coarse and gelatinous precipitates
Medium Porosity
  • Flowrate: Medium, 60mL/min.
  • Surface: Smooth
  • Particle Retention: 5 to 10μm
  • Applications: General lab uses
Fine Porosity
  • Flowrate: Slow, 5mL/min.
  • Surface: Smooth, dense texture
  • Particle Retention: 1 to 3μm
  • Applications: Excellent retention; suitable for finest particles or precipitates

Description & Specifications

Specifications

Circle