Fisherbrand™ SPE Columns
Manufacturer:  Fisherbrand™ FLP466R
 View more versions of this product
Catalog No. P466R
Description
Description
  • With Florisil and Alumina
Specifications
Specifications
Magnesium Silicate
1g
SPE Column
60Å
Magnesium Silicate
1000mg
Normal Phase
Referenced in many EPA methods, ideally suited for Pesticides and Metals
Florisil™
Florisil™
Documents