Fisherbrand™ PrepSep™ Large-Volume SPE Columns
Versatile design provides rapid flowthrough using syringe, vacuum box, or robot technology
Supplier:  Fisherbrand™ P447
 View more versions of this product
Catalog No. P447
Description
Description
  • Sorbent packings separate and retain sample analytes from their matrices; retrieve analytes by eluting with an appropriate solvent
  • Packing type is clearly marked on upper rim of each column for easy identification
  • Economically disposable; inexpensive enough to use once then throw away, eliminating cross-contamination and cleanups
  • Large-reservoir columns feature 12 to 60mL reservoirs that accept larger sample volumes; also feature a conical shape for easy filling
Specifications
Specifications
14 mL
SPE
Empty