Fisherbrand™ Pre-Cleaned Straight-Sided Jars
Certified for background contaminants
$157.26 - $184.56
Specifications
Certifications/Compliance Certificate of analysis
Closure Liner F217
Closure Material HDPE
Closure Type Screw Cap
Includes Polypropylene cap with F-217 Foam liner
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
05-719-732
View Documents Promotion Details
Fisherbrand™
05719732
500 mL 89-400 mm
Case of 24 Each for $184.56
05-719-733
View Documents Promotion Details
Fisherbrand™
05719733
1000 mL 89-400 mm
Case of 12 Each for $157.26
Description
Description
Certificate of analysis included
Environmental test, pharmaceutical and general laboratories.
Specifications
Specifications
Certificate of analysis
HDPE
Polypropylene cap with F-217 Foam liner
F217
Screw Cap