missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Pre-Cleaned Jugs
Certified for background contaminants
Manufacturer: Fisherbrand™ 05719729
Includes: Polypropylene cap with F-217 foam liner
Description
Environmental test, pharmaceutical and general laboratories.
Specifications
F217 | |
38-400 mm | |
135.2 oz., 4000mL | |
135.2 oz. | |
Plastic |
Polypropylene | |
Screw Cap | |
4000 mL | |
Polypropylene cap with F-217 Foam liner |