Fisherbrand™ Pre-Cleaned Cylinders
Certified for background contaminants
Manufacturer:  Fisherbrand™ 05719727
 View more versions of this product
Catalog No. 05-719-727
Includes: Polypropylene cap with F-217 foam liner
Description
Description
Certificate of analysis included
Environmental test, pharmaceutical and general laboratories.
Specifications
Specifications
Certificate of analysis
Polypropylene
Screw Cap
33.81 oz., 1000mL
33.81 oz.
HDPE
F217
28-410 mm
Cylindrical
1000 mL
Polypropylene cap with F-217 Foam liner