Fisherbrand™ Pre-Cleaned Boston Round Bottles
Certified for background contaminants
Manufacturer:  Fisherbrand™ 05719718
 View more versions of this product
Catalog No. 05-719-718
Includes: Polypropylene cap with F-217 foam liner
Description
Description
Certificate of analysis included
Environmental test, pharmaceutical and general laboratories.
Specifications
Specifications
Certificate of analysis
HDPE
Screw Cap
33.81 oz., 1000mL
33.81 oz.
HDPE
F217
38-430 mm
Round
1000 mL
Polypropylene cap with F-217 Foam liner