Fisherbrand™ Pre-Cleaned Boston Round Bottles
Certified for background contaminants
Manufacturer:  Fisherbrand™ 05719715
 View more versions of this product
Catalog No. 05-719-715
Includes: Polypropylene cap with F-217 foam liner
Description
Description
Certificate of analysis included
Environmental test, pharmaceutical and general laboratories.
Specifications
Specifications
Certificate of analysis
HDPE
Screw Cap
4.22 oz., 125mL
4.22 oz.
HDPE
F217
24-410 mm
Round
125 mL
Polypropylene cap with F-217 Foam liner