Fisherbrand™ Powder Funnels
Parallel stems minimize bridging of powder
Supplier: Fisherbrand™ 3007000150
Description
Parallel stems minimize bridging of powder
- Durable polypropylene
- External ribbing prevents air locks
- Autoclavable
Specifications
15cm | |
27mm | |
13.7 cm | |
30mm |
750 mL | |
Polypropylene | |
Yes |