Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Spot Plates


For observing color reactions

Manufacturer: fisherbrand™  J219

 View more versions of this product

Catalog No. FB964A

  • / Each
  • / Case of 24 Each
 

Description & Specifications

Specifications

Product Type Spot Plate
Dimensions (L x W) 3.6 x 1.2 in. (92 x 31mm)
For Use With (Application) For observing color reactions
No. of Wells 3
Color White