Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Spot Plates


For observing color reactions

Manufacturer: fisherbrand™  J220

 View more versions of this product

Catalog No. FB964B

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Product Type Spot Plate
For Use With (Application) For observing color reactions
No. of Wells 12
Dimensions (L x W) 4.4 x 3.6 in. (112 x 92mm)
Color White