Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Pestles


For hand comfort in grinding.

Manufacturer: fisherbrand™  J-208-EMD

 View more versions of this product

Catalog No. S02595

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Material Porcelain
For Use With (Equipment) 150mL Mortar
Finish Unglazed on grinding surface
For Use With (Application) For hand comfort in grinding
Length (Metric) 15.7cm
Length (English) 6.2 in.
Autoclavable Yes
Product Type Pestle