Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Pestles


For hand comfort in grinding.

Manufacturer: fisherbrand™  J-206-EMD

 View more versions of this product

Catalog No. S02593

  • / Each
  • / Case of 18 Each
 

Description & Specifications

Specifications

Product Type Pestle
Length (English) 4.49 in.
For Use With (Equipment) 50mL mortar
Length (Metric) 11.4cm
For Use With (Application) For hand comfort in grinding
Material Porcelain
Finish Unglazed on grinding surface
Autoclavable Yes