Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Combustion Boats


Chemical porcelain labware design with resistance to heat and caustic chemicals

Manufacturer: fisherbrand™  J112

 View more versions of this product

Catalog No. FB963A

  • / Pack of 12 Each
  • / Case of 6 Pack
 

Description & Specifications

Specifications

Dimensions (L x W x H) 0.6 x 0.3 x 0.15 in. (17 x 6 x 4mm)
Product Type Combustion Boat
Color White
For Use With (Application) Scientific tools used in experiments because of their resistance to heat and caustic chemicals
Material Porcelain