Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Combustion Boats


Chemical porcelain labware design with resistance to heat and caustic chemicals

Manufacturer: fisherbrand™  J115

 View more versions of this product

Catalog No. FB963D

  • / Each
  • / Case of 18 Each
 

Description & Specifications

Specifications

Product Type Combustion Boat
Dimensions (L x W x H) 3.9 x 0.7 x 0.5 in. (100 x 20 x 13mm)
For Use With (Application) Scientific tools used in experiments because of their resistance to heat and caustic chemicals
Material Porcelain
Color White