Please login to your online account to display your discounted pricing

Fisherbrand™ Porcelain Capsule-Form Crucibles


Glazed inside and out except for rim

Manufacturer: fisherbrand™  J102

 View more versions of this product

Catalog No. FB960V

  • / Pack of 6 Each
  • / Case of 8 Pack
 

Description & Specifications

Specifications

Capacity (English) 0.9 oz.
Color White
Finish Glazed
Capacity 0.9 oz., 25mL
Product Type Crucible
Material Porcelain
Capacity (Metric) 25mL