Please login to your online account to display your discounted pricing

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps


Manufacturer: fisherbrand™  W014467

Catalog No. 02-912-088

  • / Pack of 12 Each
  • / Case of 24 Pack
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Material Plastic
Closure Liner Poly/Vinyl
Color White
Description Screw cap
Material HDPE
No. per Case 12/Pk.
Quantity 288/PK.
For Use With (Equipment) Bottle
Closure Size 70/400
Liner Material Polyvinyl
Neck Finish 70-400
Closure Type screw cap