Please login to your online account to display your discounted pricing

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps


Manufacturer: fisherbrand™  W014466

Catalog No. 02-912-087

  • / Pack of 12 Each
  • / Case of 24 Pack
 

Description & Specifications

Specifications

Sterile Yes
Closure Material Plastic
Closure Size 63/400
Closure Type screw cap
Description Screw cap
Material HDPE
Liner Material Polyvinyl
Quantity 288/PK.
Certifications/Compliance ISO 9001
Closure Liner Poly/Vinyl
Color White
For Use With Bottle
Neck Finish 63-400
No. per Case 12/Pk.