Please login to your online account to display your discounted pricing

Fisherbrand™ White Plastic Screw Bottle Caps

Manufacturer: fisherbrand™  W014465

 View all options for this product

Catalog No. 02-912-086


Description & Specifications


Certifications/Compliance ISO 9001
Closure Material Plastic
Closure Liner Poly/Vinyl
Closure Size 58/400
Closure Type screw cap
Color White
Description Screw cap
For Use With Bottle
Material HDPE
Liner Material Polyvinyl
Neck Finish 58-400
No. per Case 12/Pk.
Quantity 288/PK.
Sterile Yes

1mm polyvinyl film bonded to 1mm HDPE on a #30 white pulp paper backing. Superior to plain pulp paper because it provides excellent moisture barrier. For Fisherbrand™ Boston round and wide mouth packer bottles. General purpose suitable for a wide range of applications. Good chemical resistance suitable for mild acids, alkalis, solvents, alcohols, oils and aqueous products; poor for active hydrocarbons and bleaches.