Please login to your online account to display your discounted pricing

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps


Manufacturer: fisherbrand™  W014462

Catalog No. 02-912-291

  • / Pack of 12 Each
  • / Case of 48 Pack
 

Description & Specifications

Specifications

Sterile Yes
Closure Material Plastic
Closure Size 48/400
Closure Type screw cap
Description Screw cap
Material HDPE
Liner Material Polyvinyl
Quantity 576/PK.
Color White
Neck Finish 48-400
Certifications/Compliance ISO 9001
Closure Liner Poly/Vinyl
For Use With Bottle
No. per Case 12/Pk.