Please login to your online account to display your discounted pricing

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps


Manufacturer: fisherbrand™  W014458

Catalog No. 02-912-289

  • / Case of 576 Each
 

Description & Specifications

Specifications

Closure Type screw cap
Liner Material Polyvinyl
For Use With (Equipment) Bottle
Neck Finish 43-400
Closure Size 43/400
Sterility Sterile
Material HDPE
Quantity 576/PK.
Closure Material Plastic
Description Screw cap
Color White
Certifications/Compliance ISO 9001
Closure Liner Poly/Vinyl
No. per Case 576/Cs.