Please login to your online account to display your discounted pricing

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps


Manufacturer: fisherbrand™  W014456

Catalog No. 02-912-288

  • / Case of 576 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Material Plastic
Closure Liner Poly/Vinyl
Color White
Description Screw cap
Material HDPE
No. per Case 576/Cs.
Quantity 576/PK.
Sterility Sterile
For Use With (Equipment) Bottle
Closure Size 38/400
Liner Material Polyvinyl
Neck Finish 38-400
Closure Type screw cap