missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Polyurethane Ice Buckets

For use with liquid nitrogen, dry ice, water ice, and salt solutions

Manufacturer:  Fisherbrand™ 501864F

 View more versions of this product

Catalog No. 02-591-44


 

Specifications Description & Specifications

Specifications

4.5L
Black
95°C
Ice Bucket
7.25 in.
Polyurethane
Dry ice, water ice and salt solutions
203°F
18.5cm