Fisherbrand™ Polystyrene Weighing Dishes
Ideal for weighing, dispensing or storing applications.
$34.85 - $73.48
Specifications
Shape Flat
Type Weighing Dish
Disposable Yes
For Use With (Application) Ideal for weighing, dispensing or storing applications
Height (Metric) 25.4 mm
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Price Quantity  
S67091A View Documents Fisherbrand™
08732115EM
139 mm 139 mm Case of 500 Each for $73.48
S67090A View Documents Fisherbrand™
08732113EMD
88.9 mm 88.9 mm Case of 500 Each for $34.85
Description
Description
  • Contaminant-free, antistatic and biologically inert
  • Resist dilute acids, aqueous solutions, alcohols and bases
  • Flat bottoms resist tipping
  • Made from 12mil thick polystyrene for better strength, stability and handling ease
Specifications
Specifications
Flat
Yes
25.4 mm
Weighing Dish
Ideal for weighing, dispensing or storing applications
12 mil