Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Polystyrene Jars with White Polypropylene Foamed PE-Lined Cap
Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Clear Polystyrene Jars with White Polypropylene Foamed PE-Lined Cap are available in various capacities varying from 1 oz. to 32 oz.
$77.40 - $214.03
Specifications
Closure Liner Foam Polyethylene
Closure Material Polypropylene
Autoclavable No
Closure Type Screw
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Price Quantity    
02-911-792
fisherbrand™
263983
1 oz., 30mL 43-400 Case of 72 Each for $109.00
02-912-265
fisherbrand™
263982
16 oz., 500mL 89-400 Case of 24 Each for $115.23
02-912-266
fisherbrand™
263985
32 oz., 1000mL 20-400 Case of 24 Each for $214.03
02-912-267
fisherbrand™
263984
2 oz., 60mL 53-400 Case of 48 Each for $77.40
02-912-268
fisherbrand™
263986
4 oz., 125mL 58-400 Case of 36 Each for $84.90
02-912-269
fisherbrand™
263987
8 oz., 250mL 89-400 Case of 36 Each for $129.83
Description & Specifications
Specifications
Closure Liner Foam Polyethylene
Closure Material Polypropylene
Autoclavable No
Closure Type Screw
Color Clear
Material Polystyrene
Includes Foam Polyethylene lined cap
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Rigid clear polystyrene jars offer glass-like clarity with a slip-resistant smooth surface
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.