missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Polypropylene Tubing T-Shaped Connectors

Withstand flows to 20psi.

Manufacturer:  Fisherbrand™ 227915

 View more versions of this product

Catalog No. 15-315-26B



Description

Description

  • Excellent chemical resistance
  • Polypropylene
  • Autoclavable
  •  Serrated connecting surfaces to prevent tubing slipping
  • Withstands flows to 20 p.s.i.
Specifications

Specifications

Transparent
0.218 in.
1.185 in.
Tubing Connector
20psi
0.2 in.
22.8mm
Yes
Polypropylene
30mm
6mm
5.08mm
T
0.9 in.
Documents