Please login to your online account to display your discounted pricing

Fisherbrand™ Polypropylene Sample Vials with Hinged Cap


High-density polypropylene vials

Manufacturer: fisherbrand™  M086000

 View more versions of this product

Catalog No. 03-341-76A

  • / Case of 50 Each
 

Description & Specifications

Specifications

Color Opaque
Capacity (Metric) 240mL
Closure Type Snap cap
For Use With (Application) Sample identification and organization and bar coding
Capacity 8 oz., 240mL
Capacity (English) 8 oz.
Quantity 50 Case
Material HDPE
Type Sample Vial