Please login to your online account to display your discounted pricing

Fisherbrand™ Polypropylene Sample Vials with Hinged Cap


High-density polypropylene vials

Manufacturer: fisherbrand™  M026000

 View more versions of this product

Catalog No. 03-341-75C

  • / Case of 600 Each
 

Description & Specifications

Specifications

Material HDPE
Type Sample Vial
Capacity (Metric) 60mL
For Use With (Application) Sample identification and organization and bar coding
Color Opaque
Capacity (English) 2 oz.
Closure Type Snap cap
Quantity 600 Case