Please login to your online account to display your discounted pricing
Fisherbrand™ Polypropylene Rectangular Carboys
Value your lab can rely on. Fisherbrand Polypropylene Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
$115.77 - $190.40
Specifications
Certifications/Compliance FDA, USP VI
Autoclavable Yes
Material Polypropylene
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
View More Specs
Products
Catalog Number Mfr. No. Capacity Product Type Price Quantity    
03-007-645
fisherbrand™
N3022120020
2.38 gal., 9L Carboy without spigot Case for $115.77
03-007-647
fisherbrand™
N3023210020
2.38 gal., 9L Carboy with spigot Case for $148.93
03-007-648
fisherbrand™
N3023210050
5.28 gal., 20L Carboy with spigot Case for $190.40
Description & Specifications
Specifications
Certifications/Compliance FDA, USP VI
Autoclavable Yes
Material Polypropylene
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
Shape Rectangular
Ideal for storing and dispensing lab water and reagents
  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)