Please login to your online account to display your discounted pricing
Fisherbrand™ Polypropylene Rectangular Carboys
Value your lab can rely on
Manufacturer: fisherbrand™  N3022120050
 View more versions of this product
Catalog No. 03-007-646
  • / Case
Description & Specifications
Specifications
Capacity (English) 5.28 gal.
Certifications/Compliance FDA, USP VI
Capacity (Metric) 20L
Material Polypropylene
Autoclavable Yes
Capacity 5.28 gal., 20L
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
Shape Rectangular
Product Type Carboy without spigot