missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Polypropylene Microtube Support Racks

Excellent for microbiology work or any work using microtubes

Manufacturer:  Fisherbrand™ F188400000

 View more versions of this product

Catalog No. 03-213-1



 

Specifications Description & Specifications

Specifications

4.37 in.
400μL Tubes
White
4.37 in.
22.8cm
Rack, Microtube
72.0
9 in.
4/case
11cm
11cm
No
Polypropylene
9 x 4.37 x 4.37 in. (22.8 x 11 x 11cm)
Promotions

Promotions