Fisherbrand™ Polypropylene Microtiter Tubes, Plain tubes; Nonsterile; Bagged
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Manufacturer:  Fisherbrand™ RC845TPSFIS
 View more versions of this product
Catalog No. 02-681-386
Description
Description
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.
  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Nonsterile tubes are certified RNase- and DNase-free
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL
Order Info
Packed in bulk, or in convenient 8 × 12 compact tray packs or robotic racks.
Compact Tray Packs
  • Snug, smooth- fitting cover protects samples and enables stacking
  • With paper grid for easy indentification of contents
Robotic Racks
  • Specifically designed with 96-well footprint for robotic workstations
  • One-way lid design with alphanumeric grid enables easy sample identification
  • Condensation rings inside cover minimize cross-contamination
  • Tubes fit in straight, upright position
Specifications
Specifications
Sterile
Yes
Natural
HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent
RNase, DNase, ATP, DNA, pyrogen-free
1.2mL
Polypropylene
Robotic Pack
Documents