Please login to your online account to display your discounted pricing
Fisherbrand™ Polypropylene Microtiter Tubes
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Manufacturer: fisherbrand™  RC845FTPFIS
 View more versions of this product
Catalog No. 02-681-387
  • / Pack of 960 Each
  • / Case of 5 Pack
Description & Specifications
Specifications
Product Type Frosted Microtiter Tube
Capacity (Metric) 1.2mL
Color Natural
For Use With (Application) HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent
Sterile No
Packaging Robotic Pack
Certifications/Compliance RNase, DNase-free
Autoclavable Yes
Material Polypropylene